D-(-)-Mannitol 96, powder USP

Supplier: Spectrum Chemical

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

MA165-2.5KG MA165-500GM
SPCMMA165-2.5KGEA 960 EUR
SPCMMA165-2.5KG SPCMMA165-500GM
D-(-)-Mannitol 96, powder USP
D(-)-Mannitol

Mannitol, Powder, USP.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1,49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 96% Min.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR