N-Nitroso-N-methylurea

Supplier: Spectrum Chemical

N2939-5GM
SPCMN2939-5GMEA 592 EUR
SPCMN2939-5GM
N-Nitroso-N-methylurea
N-Nitroso-N-methylurea

N-Nitroso-N-methylurea, abbreviated as NMU, is an alkylating agent and a precursor in the synthesis of diazomethane.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂H₅N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00014794
CAS Number: 684-93-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR