Polyvinyl alcohol USP

Supplier: Spectrum Chemical

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

P1180-12KGBL P1180-25KGBL P1180-50KGBL
SPCMP1180-12KGBLEA 25080 EUR
SPCMP1180-12KGBL SPCMP1180-25KGBL SPCMP1180-50KGBL
Polyvinyl alcohol USP
Polyvinyl alcohol

Polyvinyl Alcohol, USP is used to relieve dry, irritated eyes. All Spectrum Chemical USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Viscosity Method Used As Reported
pH of a 4% Solution 5.0 - 8.0
Degree of Hydrolysis 85 - 89%
Loss On Drying max. 5.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR