N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine ≥98%

Supplier: Spectrum Chemical

Synonyms: 1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol), Ethylenediamine-N,N,N',N'-tetra-2-propanol

T2016-1KG
SPCMT2016-1KGEA 419 EUR
SPCMT2016-1KG
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine ≥98%
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine
Formula: C₁₄H₃₂N₂O₄
MW: 292.42 g/mol
Density: 1,013
Storage Temperature: Ambient
MDL Number: MFCD00004534
CAS Number: 102-60-3

Order Now

Specification Test Results

Assay Approx. 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR