Vanillin NF

Supplier: Spectrum Chemical
Warning

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

VA130-12KG VA130-2.5KG VA130-50KGBL
SPCMVA130-12KGEA 3710 EUR
SPCMVA130-12KG SPCMVA130-2.5KG SPCMVA130-50KGBL
Vanillin NF
Vanillin
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285
Melting Pt: ∼82 °C
Density: 1.056 g/cm³ (20 °C)
Flash Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR