Silvers skin graft blades
Supplier: SWANN MORTON
9942
SWAN9942EA 51.9 EUR
SWAN9942
Silvers skin graft blades
Scalpels and Scalpel Assemblies Scalpel Blades
This custom designed surgical stainless steel Silver's blade has a 11/2 inch (4 cm) postage stamp size cutting edge to be used with the Silvers miniature Skin Graft Knife handles.
  • Double edged
  • Sterile, individually V.P.I. protected in metal foil packets
  • Can also be used for the debridement of wounds
Ideal for harvesting small intricate grafts from areas which would not be accessible with the larger standard knives such as the ear, dorsa of the hands or scalp.
Balení: Blades are sealed in individual foil packs.