Biological Safety Cabinets, Class II for Cytotoxics, BiOptima CYTO
Supplier: Azbil Telstar Technologies
1724077 1724079
TELR1724077EA 12804.91 EUR
TELR1724077 TELR1724079
Biological Safety Cabinets, Class II for Cytotoxics, BiOptima CYTO
Fume Hoods and Enclosures Biological Safety Cabinets
The ergonomically designed BiOptima Class II microbiological safety cabinets ensure optimum efficiency and high-level energy savings. BiOptima provides operator protection with the inflow air barrier, product protection thanks to HEPA-filtered laminar downflow in the working area and environmental protection through the exhaust HEPA filter. BiOptima Cyto is available in 120 or 180 cm (4 or 6’) widths as standard.
  • Working surface is divided into sections, allowing cleaning and even sterilisation in an autoclave
  • Control panel has international colour coding