Biological Safety Cabinets, class II, BiOptima
Supplier: Azbil Telstar Technologies
1622438 1367783 1734952 1736028
TELR1622438EA 10670.76 EUR
TELR1622438 TELR1367783 TELR1734952 TELR1736028
Biological Safety Cabinets, class II, BiOptima
Fume Hoods and Enclosures Biological Safety Cabinets
The ergonomically designed BiOptima Class II microbiological safety cabinet ensures optimum efficiency and high-level energy savings. BiOptima provides operator protection with the inflow air barrier, product protection thanks to HEPA-filtered laminar downflow in the working area and environmental protection through the exhaust HEPA filter.
  • Working surface is divided into sections, allowing cleaning and even sterilisation in an autoclave
  • Control panel has international colour code to indicate cabinet status
  • Main screen shows the level of filter clogging