Digital multimeter, testo 760-3
Supplier: Testo
0590.7603
TEST0590.7603EA 225 EUR
TEST0590.7603
Digital multimeter, testo 760-3
Electrical Meters Digital Multimeters
The testo 760-3 digital multimeter is the multitasker for all professionals in the electricity sector. It enables users to carry out both simple and demanding electrical measuring tasks more reliably than ever before. The testo 760-3 automatically detects the appropriate measurement parameter via the socket assignment and therefore prevents dangerous incorrect settings.
  • Easy, reliable work thanks to automatic measurement parameter detection
  • Auto power off saves battery life
  • Rugged meter with overmould housing
  • Comfortable, one-handed operation without any