Cleanroom vacuum cleaner, CR-1200D
Supplier: TIGER-VAC
323600 323604 110134B
TIGV323600EA 339 EUR
TIGV323600 TIGV323604 141-1414
Cleanroom vacuum cleaner, CR-1200D
Vacuum Cleaners
New generation cleanroom vacuum cleaners available in dry recovery. Designed specifically for the pharmaceutical, semiconductor and microelectronics industry.
  • Multiple stage filtration system; simple and reliable filter change
  • The recovery tank is made of Type 316 stainless steel and is fully autoclavable (up to 121 °C) including the wheels
  • Powerful suction and quiet operation
Equipped with an upstream HEPA filter and a downstream ULPA filter. HEPA filter is H14 as per EN 1822. ULPA filter is U15 as per EN 1822. The upstream HEPA protects the motorMore Product Information