Anti-VIM Mouse Monoclonal Antibody [clone: 3E9]

Supplier: US Biological
135257
USBI135257EA 655 EUR
USBI135257
Anti-VIM Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Recommended dilution: Immunofluorescence: 40 ?g/ml

Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: 3000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR