Anti-AICDA Mouse Monoclonal Antibody [clone: 10B92]

Supplier: US Biological
A0855-75N
USBIA0855-75NEA 582 EUR
USBIA0855-75N
Anti-AICDA Mouse Monoclonal Antibody [clone: 10B92]
Antibodies
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: AICDA (activation-induced cytidine deaminase)
Clonality: monoclonal
Clone: 10B92
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR