Anti-ATIC Mouse monoclonal antibody unconjugated [clone: 10B230]

Supplier: US Biological
A1378-25J1
USBIA1378-25J1EA 403 EUR
USBIA1378-25J1
Anti-ATIC Mouse monoclonal antibody unconjugated [clone: 10B230]
Antibodies
Recommended dilution: Immunochemistry: 1:50-1:500

Type: Primary
Antigen: ATIC (5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase)
Clonality: monoclonal
Clone: 10B230
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Drosophila, Human, Mouse, Rat, Xenopus laevis
Order Now

SPECIFICATIONS

Antigen symbol ATIC
Antigen name 5-Aminoimidazole-4-Carboxamide Ribonucleotide Formyltransferase/IMP Cyclohydrolase
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 10B230
Reactivity Drosophila,Human,Xenopus laevis,Rat,Mouse
Host Mouse
Isotype IgG1
Western blot Yes
Form liquid
Purification purified by Protein G chromatography
Concentration ~1 mg/ml
Storage buffer PBS, pH 7,4, 0,09% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR