Anti-Interferon Stimulating Gene 15 Rabbit Polyclonal Antibody

Supplier: US Biological
I7662-94
USBII7662-94EA 1080 EUR
USBII7662-94
Anti-Interferon Stimulating Gene 15 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:200-1:1000

Type: Primary
Antigen: Interferon Stimulating Gene 15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR