Anti-Mycoplasma gallisepticum Chicken Polyclonal Antibody

Supplier: US Biological
M9750-05-5
USBIM9750-05-5EA 755 EUR
USBIM9750-05-5
Anti-Mycoplasma gallisepticum Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Mycoplasma gallisepticum
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

SPECIFICATIONS

Antigen name Mycoplasma gallisepticum
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Chicken
Isotype IgY
ELISA Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR