Anti-T-Cell Death-Associated Gene 8 Rabbit Polyclonal Antibody

Supplier: US Biological
T2010-04B
USBIT2010-04BEA 568 EUR
USBIT2010-04B
Anti-T-Cell Death-Associated Gene 8 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: T-Cell Death-Associated Gene 8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen name T-Cell Death-Associated Gene 8
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
Amino acid number 200 to 250
Form liquid
Purification purified by immunoaffinity chromatography
Concentration ~0,5 mg/ml
Storage buffer PBS, 0,2% gelatin, 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR