UV inspection lamp, B-100AP
Supplier: UVP ULTRA VIOLET PRODUCTS
95-0127-02M 95-0127-04M 95-0127-07M
630-0716EA 1190 EUR
630-0716 UVPA95-0127-04M UVPA95-0127-07M
UV inspection lamp, B-100AP
Lights UV Lights, Lamps and Bulbs
This lamp provides 100 W of high intensity, longwave (365 nm) illumination. It can be used for quality control, industrial inspection, and non-destructive testing.
  • Lamp head (Wר: 248×152 mm) rotates 360° when attached to base, for hands free operation
  • Cool-Touch™ housing keeps lamp cool to the touch
  • Filter is recessed to prevent breakage
Informace pro objednávání: Includes. 100 W spot bulb, transformer, and lead. Replacement bulbs sold separately.