Stericlin sterilisation pouches, flat, paper 70g/m with see-through film, with indicators for STEAM, EO and FORM, 15 x 50 cm, DIN EN ISO 11607 1 * 1.000 items
Supplier: VP Medical Packaging
Stericlin sterilisation pouches, flat, paper 70g/m with see-through film, with indicators for STEAM, EO and FORM, 15 x 50 cm, DIN EN ISO 11607 1 * 1.000 items