Conical stoppers, Versilic®
Supplier: Saint Gobain Life Sciences
263070
VERN263070EA 42.6 EUR
VERN263070
Conical stoppers, Versilic®
Stoppers Silicone Stoppers
These silicone stoppers are non toxic.
  • Biologically neutral, good chemical resistance
  • Resistant to temperatures from −60 to +200 °C (up to +220 °C for short periods)
  • Resistant to ageing, ozone and repeated sterilisation
Suitable for use with foodstuffs in accordance with FDA.
DIN 12781
Certifikace: DIN 12781