Tube clamps, Pure-Fit® TCL
Supplier: Saint Gobain Life Sciences
PFTCL150 PFTCL250
VERNPFTCL150EA 2270 EUR
VERNPFTCL150 VERNPFTCL250
Tube clamps, Pure-Fit® TCL
Clamps Tubing and Sanitary Clamps
Pure-Fit® TCL large tube clamps provide the same innovative clamping technology as Pure-Fit TC but are designed to accommodate large diameter and thick wall tubing applications.
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Custom colours and identification available
  • Fully autoclavable and sterilisable
Pure-Fit TCL is compatible with standard, braided (reinforced) and multilayer tubing. In addition to providingMore Product Information