BOD bottles, WHEATON®
Supplier: DWK Life Sciences
227497-01 227497-00G 227497-00 227494-00 227494-01 227498 227497-03G 227494-02G 227494-03 227494-03G 227494-99 227494-99G 227497-01G 227497-02 227497-03 227497-04 227497-04G 227497-05 227497-05G 227497-06 227497-06G 227497-07 227497-07G 227497-08 227497-02G 227494-02 227494-00G 227497-09G 227497-08G 227497-15 227497-10 227497-10G 227497-11 227497-11G 227497-12 227497-12G 227497-13 227497-13G 227497-14G 227497-09 227497-15G 227497-16 227497-16G 227497-17 227497-17G 227497-18 227497-18G 227497-14
WHEA227497-01EA 822 EUR
WHEA227497-01 WHEA227497-00G WHEA227497-00 WHEA227494-00 WHEA227494-01 WHEA227498 WHEA227497-03G WHEA227494-02G WHEA227494-03 WHEA227494-03G WHEA227494-99 WHEA227494-99G WHEA227497-01G WHEA227497-02 WHEA227497-03 WHEA227497-04 WHEA227497-04G WHEA227497-05 WHEA227497-05G WHEA227497-06 WHEA227497-06G WHEA227497-07 WHEA227497-07G WHEA227497-08 WHEA227497-02G WHEA227494-02 WHEA227494-00G WHEA227497-09G WHEA227497-08G WHEA227497-15 WHEA227497-10 WHEA227497-10G WHEA227497-11 WHEA227497-11G WHEA227497-12 WHEA227497-12G WHEA227497-13 WHEA227497-13G WHEA227497-14G WHEA227497-09 WHEA227497-15G WHEA227497-16 WHEA227497-16G WHEA227497-17 WHEA227497-17G WHEA227497-18 WHEA227497-18G WHEA227497-14
BOD bottles, WHEATON®
Bottles BOD Bottles
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area