Stopcocks, needle-valve, angled
Supplier: witeg Labortechnik
0.243.250
WITG0.243.250EA 24.4 EUR
WITG0.243.250
Stopcocks, needle-valve, angled
Stopcocks
Needle valve stopcock, Duran, with PTFE-needle valve, sealing system and overtwist safety device.
  • Very good chemical resistance
  • Very good temperature resistance