Adapters, Cone without Stopcock
Supplier: witeg Labortechnik
1.681.029
WITG1.681.029EA 15.2 EUR
WITG1.681.029
Adapters, Cone without Stopcock
Adapters Adapters, Joints with Tube Connectors
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Suction tube, with cone and without stopcock, bent.