Recirculating Air Filter System With Exhaust Air Monitoring
Supplier: asecos
39306 40259
135-2879EA 1746 EUR
135-2879 MSGA40259
Recirculating Air Filter System With Exhaust Air Monitoring
Air Purifiers
Recirculating air filter systems – avoid complex wall breakthroughs and expensive exhaust air ducts.
  • High flexibility in choosing the installation location
  • Active personal protection through safe capture of harmful vapours with retention in the filter system
  • Very low noise, only approx. 23 dB (A)
  • Housing made of sheet-steel powder-coated
  • With multi-stage filter system, secondary filter, connection cable and mains plug
  • Includes monitoring electronics for exhaust air and filter monitoring and display for status and error indication
  • Key control panel for
More Product Information
Order Now
SPECIFICATIONS
Airflow rate 10 m³/h
Environmentally Preferable
Electrical 85 - 265 V, 50/60 Hz, 0,13 A (max. current consumption)
Noise level 23 dB