Viton Transfer Pump Tubing, Viton®, 1/8" ID x 3/16" OD; 25 ft
Supplier: Avantor Fluid Handling
keep light sensitive samples safe.
- Global market leader in range of formulations for most extensive chemical compatibility
Applications: Acid and solvent transfer; high-temperature work.<br />Characteristics: Opaque black.<br />Certification: Tubing is manufactured according to GMP. <br />Temperature range: –25 to 399 °F (–32 to 204 °C)<br />Sterilize: Not recommended.