Masterflex® L/S® Replacement Motor for Digital Benchtop Modular Drive with Remote I/O, 100 rpm
Supplier: Avantor Fluid Handling
VWR Item#MFLX07557-12
Your Organization's Price:
Make the most of your Masterflex drives replacement parts and other accessories.
  • Extend the life of your Masterflex pump with these replacement parts
  • Suitable for use with 07557-10 benchtop modular drive
Choose replacement motors based on your Masterflex Modular System.