Masterflex® Fitting, Polycarbonate, Two Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
Supplier: Avantor Fluid Handling
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Two ports with female luer locks
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
<B>Warning:</B> This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.