Micropump GJ Series B-Mount Cavity Gear Pump Head, SS/PPS/PTFE, Internal Bypass; 0.64 mL/rev
Supplier: Avantor Fluid Handling
VWR Item#MFLX74004-05
Your Organization's Price:
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.