Bev-A-Line Transfer Tubing, Bev-A-Line® XX, 3/8" ID x 1/2" OD; 250 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX06491-56
Your Organization's Price:
Particulate will not adhere to liner.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: High-purity applications and air sampling. Ideal for particulate matter (will not adhere to liner) and applications requiring UV resistance.<br />Characteristics: HYTREL® polyester liner and UV resistant PVC. Tubing is coextruded for improved flexibility. Shell is opaque black.<br />Certification: Liner resin is NSF-approved for potable water.<br />Temperature range: –20 to 160 °F (–51 to 71 °C)<br />Sterilize: Ethylene oxide only