Masterflex® Inert in-line inlet check valve; 0.014 Cv, PEEK/EPDM
Supplier: Avantor Fluid Handling
No maintenance required!
- Ideal for high-purity, low pressure
- No metal components - inert flow path
- No back flow leakage with back pressure of 0 to 100 psi
- 1/4-28 UNF threaded connections
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.