Masterflex® Inert in-line female check valve; 0.014 Cv, PEEK/Chemraz
Supplier: Avantor Fluid Handling
VWR Item#MFLX01355-28
Your Organization's Price:
No maintenance required!
  • Ideal for high-purity, low pressure
  • No metal components - inert flow path
  • No back flow leakage with back pressure of 0 to 100 psi
  • 1/4-28 UNF threaded connections
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.