Hand-operated acid-transfer pump, 6 strokes/gallon, female buttress adapter
Supplier: Avantor Fluid Handling
VWR Item#MFLX06433-40
Your Organization's Price:
Ideal for most acid/base or ionic salt applications.
  • UL-listed for sulfuric acid transfer at 50% concentration
  • Withstand temperatures up to 120 °F (49 °C)
  • Self-priming, will not siphon
These plastic hand-operated drum pumps are ideal for heavy-duty applications that require acid-transfer. All models are constructed of solvent-welded PVC body and shaft with Viton® valves and seals and feature 6 strokes/gallon (2 strokes/liter).