Bev-A-Line Transfer Tubing, Bev-A-Line® V, 3/8" ID x 1/2" OD; 50 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX06491-17
Your Organization's Price:
Needs no connectors—heat-bondable.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: Safe for food, beverage, drug, lab, and general use—can withstand higher temperatures than Bev-A-Line® IV tubing. Safe for most alcohols; avoid strong solvents.<br />Characteristics: Polyethylene liner cross-linked to ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Needs no connectors—heat-bondable. Translucent. <br />Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements.<br />Temperature range: –60 to 200 °F (–51 to 93 °C)<br />Sterilize: With autoclave, ethylene oxide, or dry heat.