Bev-A-Line Transfer Tubing, Bev-A-Line® XX, 11/64" ID x 1/4" OD; 500 Ft
Supplier: Avantor Fluid Handling
Particulate will not adhere to liner.
- Ideal for general-purpose, single-use, and extended fluid path applications
Applications: High-purity applications and air sampling. Ideal for particulate matter (will not adhere to liner) and applications requiring UV resistance.<br />Characteristics: HYTREL® polyester liner and UV resistant PVC. Tubing is coextruded for improved flexibility. Shell is opaque black.<br />Certification: Liner resin is NSF-approved for potable water.<br />Temperature range: –20 to 160 °F (–51 to 71 °C)<br />Sterilize: Ethylene oxide only