Hand-Operated Rotary Drum Pump, SS, 10 GPM, 40"L Tube, 5000 SSU
Supplier: Avantor Fluid Handling
VWR Item#MFLX06510-10
Your Organization's Price:
Dispense a variety of liquids compatible with polypropylene, Ryton® PPS, cast iron, aluminum, stainless steel, or PTFE.
  • Lightweight and easy to use
  • Smooth, non-pulsating flow
  • Operate either in forward or reverse direction with a discharge hose
Polypropylene rotary pump (06510-00) is excellent for most organic acids, waxes, plating solutions, detergents and water soluble inorganic chemicals. Ryton PPS rotary pump (06510-05) is able to dispense aggressive chemicals such as esters, ketones and hydrocarbons. Ryton PPS pumps include a stainless steel discharge tube with a 3-piece stainless steel suction tube. Stainless steel pump (06510-10) with PTFE seal is suitable for pumping chlorinated or petroleum-based solvents. For larger flows with compatible liquids, there are now cast iron (06510-17) and aluminum (06510-19) pumps.<br /><br />Call our Application Specialists to order optional 1" PVC 5-ft tubing with hose clamps.
<b>Delivery information: </b>Adapter type 11/2" x 2" NPT bung.