GF Signet Paddlewheel Flow Sensor, 4-20 mA Output, PVDF, 0.5" to 4" Pipe Size
Supplier: Avantor Fluid Handling
VWR Item#MFLX32512-17
Your Organization's Price:
Easily configure to your needs.
  • Built-in user interface for on-site configuration
  • Low power and high resolution
Fluid measurement with paddlewheel sensors<br />Digital (S3L) or 4 to 20mA outputs, or Flow Switch, or Pulse output (multi-function)<br />Allows for up to six sensors to Signet 8900 Controller<br />Low flow capabilities down to 0.1 m/s (0.3 ft/s)<br />Polypropylene or PVDF wetted materials