Kodiak Digital Ultrasonic Drum Level Gauge; Battery Powered
Supplier: Avantor Fluid Handling
Never come up empty—reads to ±0.50 gallon (1 liter).
- Resists temperature fluctuations
- Ideal for applications involving diesel fuel, oils, hydraulic fluid, and water†
- Works with 15, 30, or 55 gallon (57, 114, or 208 liter) vertical drums
- Digital display indicates volume in gallons or liters numerically and as an analog bar graph
Ultrasonic drum level gauges feature all laser-welded design and custom integrated signal conditioning for calibration and temperature compensation.
Connects directly to the 3/4" bunghole of a drum, or the threaded adapter may be removed to reveal a 2" thread as well. Digital display indicates volume in gallons or liters numerically and as an analog bar graph. A red, low-level warning light also illuminates when total volume drops below 10%. <br />Operates on two 9 V batteries (not included) to display a static reading; update the level readings by pushing the "Refresh/Select" button. Auto shut-off feature prolongs battery life. <br />†Not intended for use with gasoline.