Masterflex® Ismatec® Pump Tubing, 2-Stop, Platinum-Cured Silicone, 2.29 mm ID; 6/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX95602-44
Your Organization's Price:
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • No leachable additives or plasticizers
  • Inner diameter: 2.29 mm
Appearance: Opaque<br />Temperature: -70 to 52 °C<br />Life at 100 rpm: 1000 hrs<br /><br />The overall length of platinum-cured silicone 95602-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.<br />Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.