Fitting, Polypropylene, Y-Connector, Hose Barb Union, 6 mm ID; 100/Pk
Supplier: Avantor Fluid Handling
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
- Connection Type: 6 mm ID; 100/PK
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.<br />Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.