Bloodstopper, 4-in-1
Supplier: ORKLA CARE
1910 1911
118-0018EA 13.6 EUR
118-0018 118-0022
Bloodstopper, 4-in-1
First Aid Supplies Wound Dressings
This sterile universal dressing consists of two elastic bandages (mini version only contains one bandage) and one compress with a non stick wound contact layer. The compress is stitched to one of the bandages. The dressing is wrapped in a pouch with printed pictorial user instructions. The Bloodstopper is to be used in first aid situations. It has four functions.
  • Pressure pad for heavy bleeding
  • Protective dressing for superficial wounds
  • Burn dressing with special wound-protecting surface
  • Temporary support dressing
The versatile Bloodstopper canMore Product Information