Portable refractometers, PAL series
Supplier: Atago
4489 4492 4421 4423 4424 4436 4437 4438 4440 4441 4443 4444 4445 4446 4447 4448 4449 4450 4452 4456 4457 4458 4459 4460 4461 4462 4463 4467 4469 4485 4502
635-0159EA 339 EUR
635-0159 635-0161 635-0229 635-0230 635-0231 635-0240 635-0241 635-0242 635-0244 635-0245 635-0247 635-0248 635-0249 635-0250 635-0251 635-0252 635-0253 635-0254 635-0256 635-0260 635-0261 635-0262 635-0263 635-0264 635-0265 635-0266 635-0267 635-0271 635-0273 635-0280 ATAG4502
Portable refractometers, PAL series
Refractometers Handheld Refractometers
The PAL series with special scales are pocket instruments delivering rapid results for automotive, aviation, heat transfer and industrial applications.
Certifikace: IP 65 allows rinsing off the sample with running water.