Carboys, narrow neck, with screw cap, HDPE
Supplier: VITLAB
81660 81662
VITL81660EA 0 EUR
VITL81660 VITL81662
Carboys, narrow neck, with screw cap, HDPE
Bottles Carboys
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
With stable carrying handle and screw cap. The 25 and 50 L sizes come equipped with two carrying handles. Complete with exchangeable. easily operated tap made from PP with a ³⁄₄" pipe fitting.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904