Ethyl-3-phenylglycidate (mixture of cis and trans isomers), Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethyl 3-phenylglycidate
M-Clarity™ quality level = MQ300
W245402-SAMPLE W245402-1KG W245402-5KG
SIALW245402-SAMPLEEA 54.3 EUR
SIALW245402-SAMPLE SIALW245402-1KG SIALW245402-5KG
Ethyl-3-phenylglycidate (mixture of cis and trans isomers), Sigma-Aldrich®
Ethyl 3-phenylglycidate (mixture of cis and trans isomers)
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
MDL Number: MFCD00005123
CAS Number: 121-39-1
EINECS: 204-467-3