Chlordane

Supplier: EHRENSTORFER GMBH

EHERC11200000EA 49.6 EUR
EHERC11200000
Chlordane
Chlordane
Formula: C₁₀H₆Cl₈
CAS Number: 57-74-9
UN: 2996
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR