Gallium indium alloy (75.5:24.5 w%), eutectic ≥99.99% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Gallium-indium eutectic
12478.03
12478.09
12478.18
12478.03EA
44.8
EUR
12478.03
12478.09
12478.18
Gallium indium alloy (75.5:24.5 w%), eutectic ≥99.99% (metals basis)
Gallium Indium alloy (75.5:24.5 w%), eutectic
Purity: 99.99%
Molecular Formula: Ga:In; 75.5:24.5 wt%
Physical Form: Liquid
Density: 6.2
Appearance: Silver
Melting Point: 15.7°
UN No.: 1760
MDL No.: MFCD00144387
Melting Pt:
15.7 °C Density: 6.2 Storage Temperature: Ambient |
MDL Number:
MFCD00144387 CAS Number: 56451-02-6 EINECS: 231-163-8 UN: 1760 ADR: 8,III |