Gallium indium alloy (75.5:24.5 w%), eutectic, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Gallium-indium eutectic
M-Clarity™ quality level = MQ100
495425-25G
495425-5G
SIAL495425-25GEA
379
EUR
SIAL495425-25G
SIAL495425-5G
Gallium indium alloy (75.5:24.5 w%), eutectic, Sigma-Aldrich®
Gallium Indium alloy (75.5:24.5 w%), eutectic
Storage Temperature:
Ambient |
MDL Number:
MFCD00144387 CAS Number: 56451-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...