2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
Supplier: Thermo Scientific
Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium
SB00727.1 SB00727.10 SB00727.50
MAYBSB00727.1EA 0 EUR
MAYBSB00727.1 MAYBSB00727.10 MAYBSB00727.50
2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
2-Mercaptopyridine N-oxide sodium salt
Formula: C₅H₄NNaOS
MW: 149.15 g/mol
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5