(±)-1-Benzyl-3-hydroxypiperidine hydrochloride

Supplier: Thermo Scientific

Synonyms: 1-Benzyl-3-piperidinol hydrochloride

AC13425DA AC13425EA AC13425.50
MAYBAC13425DAEA 0 EUR
MAYBAC13425DA MAYBAC13425EA MAYBAC13425.50
(±)-1-Benzyl-3-hydroxypiperidine hydrochloride
(±)-1-Benzyl-3-hydroxypiperidine hydrochloride
Formula: C₁₂H₁₈ClNO
MW: 227.7304 g/mol
MDL Number: MFCD00044200
CAS Number: 105973-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR