Iminostilbene 97%
Supplier: Thermo Fisher Scientific
Synonyms: 5H-Dibenzo[b,f]azepine
162540100 162541000
ACRO162540100EA 42.8 EUR
ACRO162540100 ACRO162541000
Iminostilbene 97%
Iminostilbene
Formula: C₁₄H₁₁N
Melting Pt: 197…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00005071
CAS Number: 256-96-2
EINECS: 205-970-0
Order Now
Specification Test Results
Appearance Yellow to orange Powder
Infrared spectrum Conforms
Melting point 196 to 202 °C
GC ≥96.0 %