Iminostilbene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5H-Dibenzo[b,f]azepine

M-Clarity™ quality level = MQ200

143650-1G 143650-5G
SIAL143650-1GEA 61 EUR
SIAL143650-1G SIAL143650-5G
Iminostilbene, Sigma-Aldrich®
Iminostilbene
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
MDL Number: MFCD00005071
CAS Number: 256-96-2
EINECS: 205-970-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR